VWR® Sharps Container Systems, Red
2.5 gal., Non-Stackable, 25.4W×15.2D×38.1H cm (10W×6D×15"H), Red
  19001-026
VWR® Sharps Container Systems, Red
  19001-026
 :  
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red
2.5 gal., Non-Stackable, 25.4W×15.2D×38.1H cm (10W×6D×15"H), Red
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.
  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The stackable sharps containers are puncture-resistant and both autoclaves-safe and incinerator-safe. The tortuous path systems restrict hand entry. The extended neck sharps containers feature a funneled vertical drop and extended handles.
  • Capacité:
    2.5 gal.
  • Color:
    Red
  • Dimensions:
    25.4W×15.2D×38.1H cm (10W×6D×15"H)
  • Style:
    Non-Stackable