WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
15 mm, Orange
Catalog # 16300-085
Supplier: DWK Life Sciences (KIMBLE)
: 242423
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Catalog # 16300-085
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).
Specifications
- Taille du bouchon:15 mm
- Color:Orange