WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences | Avantor
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
13 mm, Purple
Catalog # 16300-077
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Catalog # 16300-077
Supplier:  
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
13 mm, Purple
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
DROPPER TIP CAP PP 13MM CS100 Catalog #16300-141
Product Details & Documents
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).
Spécifications
  • Taille du bouchon:
    13 mm
  • Color:
    Purple