Polypropylene Snap-Seal™ Sample Containers, Corning®
240 ml (8.1 oz.), Snap-Seal™ Sample Containers
Supplier: Corning
Fournisseur #: 1730-8
Jump to:
Product Details & Documents
PP, disposable, translucent.
- Snap-Seal locking device keeps the cap closed and secure
- Hinged cap reduces the chance of sample contamination
- Graduations in both millilitres and ounces
- Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
Spécifications
- Capacité:240 ml (8.1 oz.)
- Désignation:Snap-Seal™ Sample Containers
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



