Adjustable Wrist Grounding Strap, STATICO
Adjustable Wrist Grounding Strap, STATICO
Wrist strap (band) only, —
Catalog # 14223-712
Supplier:   Statico
Supplier #:  S0101B
Adjustable Wrist Grounding Strap, STATICO
Catalog # 14223-712
Supplier:  Statico
Adjustable Wrist Grounding Strap, STATICO
Adjustable Wrist Grounding Strap, STATICO
Wrist strap (band) only, —
Price
$53.06
Log in for availability
Each Pack of 20
Product Details & Documents
Adjustable light blue wrist grounding strap with 360⁰ skin contact
A 1 Meg current limiting resistor is molded-in at the snap end termination of the cord
Certifications: Meets ANSI/ESD S1.1-2013 & ANSI/ESD S20.20-2014.
Ordering information: Available with either 1.8 m (6') or 3.6 m (12') Coil Cord and an Alligator Clip.
Specifications
  • Désignation:
    Wrist strap (band) only
  • Cord Length:
Recommendations