Adjustable Wrist Grounding Strap, STATICO
Wrist strap with coil cord and alligator clip, 1.8 m (6')
14223-708
: Statico
: S0101
Adjustable light blue wrist grounding strap with 360⁰ skin contact
A 1 Meg current limiting resistor is molded-in at the snap end termination of the cord
: Meets ANSI/ESD S1.1-2013 & ANSI/ESD S20.20-2014.
: Available with either 1.8 m (6') or 3.6 m (12') Coil Cord and an Alligator Clip.
- Désignation:Wrist strap with coil cord and alligator clip
- Cord Length:1.8 m (6')