Pipette Tips, Sorenson BioScience
QuickRack, 1 - 200 µl, Naturel
14214-395
: 30430
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
- Volume:1 - 200 µl
- Tip Color:Naturel
- Tip Style:QuickRack