Pipette Tips, Sorenson BioScience
MicroReach, 0.1 - 10 µl, Naturel
Catalog # 14006-902
Supplier: Sorenson Bioscience
Fournisseur #: 23580
Jump to:
Product Details & Documents
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Spécifications
- Volume:0.1 - 10 µl
- Tip Color:Naturel
- Tip Style:MicroReach