VWR®, Beaker Cups, Polystyrene
50 ml (1.7 oz.)
  13916-015
VWR®, Beaker Cups, Polystyrene
  13916-015
VWR®, Beaker Cups, Polystyrene
VWR®, Beaker Cups, Polystyrene
50 ml (1.7 oz.)
$363.34
Case of 500
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
  • Disposable
  • Polystyrene fabrication
  • Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
  • Capacité:
    50 ml (1.7 oz.)