VWR®, Beaker Cups, Polystyrene
50 ml (1.7 oz.)
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
- Disposable
- Polystyrene fabrication
- Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
- Capacité:50 ml (1.7 oz.)