VWR® Bath Fluids
PolyScience polytherm S150, 3.8 l (1 gal.), VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems, 50...150 °C (122...302 °F), (25°C) 50 cSt
Catalog # 13270-794
Supplier: VWR International
Jump to:
Product Details & Documents
These heat-transfer fluids provide superior thermal stability for high or low temperatures.
- Silicone heat transfer fluid
Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.
Packaging: Bottles are packaged in a carton for shipment.
Spécifications
Spécifications
- Désignation:PolyScience polytherm S150
- Taille:3.8 l (1 gal.)
- Temperature Range:50...150 °C (122...302 °F)
- For Use With:VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems
- Capacité de flacon:(25°C) 50 cSt
Spécifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



