Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Soft-Wall Panels, Trax | Avantor

Soft-Wall Panels, Trax

10 ft., 8 ft.
Catalog # 12999-006
Fournisseur #:  SWP20ST108
Soft-Wall Panels, Trax
Catalog # 12999-006
Soft-Wall Panels, Trax

10 ft., 8 ft.

Price
1 737,40 $
Log in pour la disponibilité
Each
Product Details & Documents

These standard softwall panels are ideal for forming walls of a clean enclosure or for directing air flow under a laminar flow hood

Dual-Lock™ attachment system and bottom weights are included with each panel. Shipping weights vary depending on size. Thickness: 20mil.

Spécifications
  • Longueur:
    8 ft.
  • Width:
    10 ft.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.