Soft-Wall Panels, Trax | Avantor
Soft-Wall Panels, Trax
6 ft., 4 ft.
Catalog # 12999-002
Fournisseur #:  SWP20ST64
Soft-Wall Panels, Trax
Catalog # 12999-002
Soft-Wall Panels, Trax
6 ft., 4 ft.
Price
$723.93
Log in pour la disponibilité
Each
Product Details & Documents
These standard softwall panels are ideal for forming walls of a clean enclosure or for directing air flow under a laminar flow hood
Dual-Lock™ attachment system and bottom weights are included with each panel. Shipping weights vary depending on size. Thickness: 20mil.
Spécifications
  • Longueur:
    4 ft.
  • Width:
    6 ft.