Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

VWR® Modular Heating Blocks for Vials

23 mm Vial Block, Vials, Ext.Ø 23 mm, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2"), 8
Catalog # 12621-134
Supplier:   VWR International
VWR® Modular Heating Blocks for Vials
Catalog # 12621-134
Supplier:  VWR International
VWR® Modular Heating Blocks for Vials
VWR® Modular Heating Blocks for Vials

23 mm Vial Block, Vials, Ext.Ø 23 mm, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2"), 8

Price
450,54 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Designed for sample/serum and scintillation vials, these units are constructed from a solid anodized aluminum block.

  • Now with Best-in-Class, Exclusive 5-Year Warranty

The close contact of tubes to block walls allows for maximum heat retention. Single block size.

Specifications
  • Désignation:
    23 mm Vial Block
  • For Vials:
    Ext.Ø 23 mm
  • L×W×H:
    9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
  • For Use With:
    Vials
  • No. of Wells:
    8
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.