VWR® Modular Heating Blocks for Vials
21 mm Vial Block, Vials, Ext.Ø 21 mm, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2"), 9
Catalog # 12621-132
Supplier: VWR International
Jump to:
Product Details & Documents
Designed for sample/serum and scintillation vials, these units are constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.
Specifications
- Désignation:21 mm Vial Block
- For Vials:Ext.Ø 21 mm
- L×W×H:9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
- For Use With:Vials
- No. of Wells:9
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



