Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Quick-Dry Foam Squeegees, Perfex | Avantor

Quick-Dry Foam Squeegees, Perfex

Foam Squeegee Blade, 61 cm (24")
Catalog # 11311-118
Supplier:   Perfex
Fournisseur #:  9724
Quick-Dry Foam Squeegees, Perfex
Catalog # 11311-118
Supplier:  Perfex
Quick-Dry Foam Squeegees, Perfex
Quick-Dry Foam Squeegees, Perfex

Foam Squeegee Blade, 61 cm (24")

Price
$150.89
Log in pour la disponibilité
Each Case of 10
Product Details & Documents

Twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces

Note: Not recommended for abrasive surfaces.

Feature a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, nonmarring, and easy to clean with warm water. Resistant to water, oil, grease, detergents, sanitizers, and solvents.

Ordering information: Polymer adjustable handle sold separately.

Spécifications
  • Désignation:
    Foam Squeegee Blade
  • Longueur:
    61 cm (24")
Accessories

Informations sur le produit

  • Catalog #
    Handle Material
    Handle Color
    Availability
    Price
  • Adjustable Handle with Long Connector
    Adjustable Handle with Long Connector
    53200-100
    Polymer
    White
    Log in pour la disponibilité
    Each - $141.82

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.