Disintegration Testing, Quality Lab Accessories
Clear Glass Disintegration Beaker with Flared Top, 900 mL
Catalog # 11010-348
Supplier: QLA
Fournisseur #: GLA900-DB
Jump to:
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Spécifications
- Désignation:Clear Glass Disintegration Beaker with Flared Top, 900 mL
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.