Mod-Rack™ Pegboard, Interdyne
30", 30", 50
Catalog # VF9-V3030
Supplier: Interdyne
: V3030
Jump to:
Product Details & Documents
Stainless steel pegboard features interchangeable white polypropylene pegs, integral drip trough, and clear plastic drain tube
Completely autoclavable and stain-resistant. Wall bracket included.
: Accessories including a funnel rack, drain basket, screen insert, and pipet rack are also available; contact your Avantor sales representative for more information.
Spécifications
- Largeur:30"
- Hauteur:30"
- No. of Pegs:50
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Screen Insert for Drain Trough, Stainless SteelScreen Insert for Drain Trough, Stainless Steel97034-242Log in pour la disponibilitéEach - $188.84
Informations sur le produit
- Catalog #AvailabilityPrice
- Screen Insert for Drain Trough, Stainless SteelScreen Insert for Drain Trough, Stainless Steel97034-244Log in pour la disponibilitéEach - $199.54
Informations sur le produit
- Catalog #AvailabilityPrice
- Drain Basket for Small Glassware Items
Informations sur le produit
- Catalog #AvailabilityPrice
- Petite C-Fold Paper Towel Dispenser
Informations sur le produit
- Catalog #AvailabilityPrice
- Vertical Liquid Soap Dispenser