Mod-Rack™ Pegboard, Interdyne
30", 30", 50
Catalog # VF9-V3030
Supplier:   Interdyne
 :  V3030
Mod-Rack™ Pegboard, Interdyne
Catalog # VF9-V3030
Supplier:  Interdyne
Mod-Rack™ Pegboard, Interdyne
Mod-Rack™ Pegboard, Interdyne
30", 30", 50
Price
$1,624.21
Log in pour la disponibilité
Each
Product Details & Documents
Stainless steel pegboard features interchangeable white polypropylene pegs, integral drip trough, and clear plastic drain tube
Completely autoclavable and stain-resistant. Wall bracket included.
 : Accessories including a funnel rack, drain basket, screen insert, and pipet rack are also available; contact your Avantor sales representative for more information.
Spécifications
  • Largeur:
    30"
  • Hauteur:
    30"
  • No. of Pegs:
    50
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Screen Insert for Drain Trough, Stainless Steel
    Screen Insert for Drain Trough, Stainless Steel
    97034-242
    Log in pour la disponibilité
    Each - $188.84
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Screen Insert for Drain Trough, Stainless Steel
    Screen Insert for Drain Trough, Stainless Steel
    97034-244
    Log in pour la disponibilité
    Each - $199.54
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Drain Basket for Small Glassware Items
    Drain Basket for Small Glassware Items
    97034-246
    Log in pour la disponibilité
    Each - $73.23
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Petite C-Fold Paper Towel Dispenser
    Petite C-Fold Paper Towel Dispenser
    97034-248
    Log in pour la disponibilité
    Each - $340.12
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vertical Liquid Soap Dispenser
    Vertical Liquid Soap Dispenser
    97034-250
    Log in pour la disponibilité
    Each - $292.85