Mod-Rack™ Pegboard, Interdyne
24", 18", 15
Catalog # VF9-V1824
Supplier: Interdyne
Supplier #: V1824
Jump to:
Product Details & Documents
Stainless steel pegboard features interchangeable white polypropylene pegs, integral drip trough, and clear plastic drain tube
Completely autoclavable and stain-resistant. Wall bracket included.
Ordering information: Accessories including a funnel rack, drain basket, screen insert, and pipet rack are also available; contact your Avantor sales representative for more information.
Spécifications
- Largeur:18"
- Hauteur:24"
- No. of Pegs:15
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Screen Insert for Drain Trough, Stainless SteelScreen Insert for Drain Trough, Stainless Steel97034-242Log in pour la disponibilitéEach - $188.84
Informations sur le produit
- Catalog #AvailabilityPrice
- Screen Insert for Drain Trough, Stainless SteelScreen Insert for Drain Trough, Stainless Steel97034-244Log in pour la disponibilitéEach - $199.54
Informations sur le produit
- Catalog #AvailabilityPrice
- Drain Basket for Small Glassware Items
Informations sur le produit
- Catalog #AvailabilityPrice
- Petite C-Fold Paper Towel Dispenser
Informations sur le produit
- Catalog #AvailabilityPrice
- Vertical Liquid Soap Dispenser
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



