Evolution® High-Column, Double-Sided Workstations, Kewaunee®
Product Details & Documents
Starter workbenches feature two front radius edge work surfaces that share two supporting columns with bases and glides.
- Work surface material is available in black epoxy resin
- ESD almond static-dissipative laminate with grounding kit
- Wallaby plastic laminate
Two support columns, a front apron, a modesty panel, and all hardware are included. Add-on workstations include all items noted above except one supporting column and base; these workstations share the column and base from an existing starter workbench.
Ordering information: Starter workbenches and add-on workstations do not include a work surface. Work surfaces are available separately. A starter workbench is required before purchasing an add-on workstation. Standard color is light neutral with a black accent strip; for other color options, contact your Avantor sales representative.
Spécifications
- Désignation:Almond ESD plastic laminate work surface, with grounding kit
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.