Canonical URL fetch failed:ERROR
Evolution® High-Column, Double-Sided Workstations, Kewaunee®
Wallaby plastic laminate work surface
  VC2460WA
Evolution® High-Column, Double-Sided Workstations, Kewaunee®
  VC2460WA
 :  
Evolution® High-Column, Double-Sided Workstations, Kewaunee®
Evolution® High-Column, Double-Sided Workstations, Kewaunee®
Wallaby plastic laminate work surface
Starter workbenches feature two front radius edge work surfaces that share two supporting columns with bases and glides.
  • Work surface material is available in black epoxy resin
  • ESD almond static-dissipative laminate with grounding kit
  • Wallaby plastic laminate
Two support columns, a front apron, a modesty panel, and all hardware are included. Add-on workstations include all items noted above except one supporting column and base; these workstations share the column and base from an existing starter workbench.
 : Starter workbenches and add-on workstations do not include a work surface. Work surfaces are available separately. A starter workbench is required before purchasing an add-on workstation. Standard color is light neutral with a black accent strip; for other color options, contact your Avantor sales representative.
  • Désignation:
    Wallaby plastic laminate work surface