Evolution® Medium-Height Workstations, Kewaunee® | Avantor
Evolution® Medium-Height Workstations, Kewaunee®
Add-on workstation, 36", 60×30"
Catalog # V407318NTK
Supplier:   Kewaunee
Fournisseur #:  V407318NTK
Evolution® Medium-Height Workstations, Kewaunee®
Catalog # V407318NTK
Supplier:  Kewaunee
Evolution® Medium-Height Workstations, Kewaunee®
Evolution® Medium-Height Workstations, Kewaunee®
Add-on workstation, 36", 60×30"
Price
$2,574.64
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
With a maximum height of 91.4 cm (36"), the starter workbenches are perfect for technical work.
  • Features a front radius edge worksurface
Starter workbenches come with two support columns, a front apron, a modesty panel, and all hardware. Add-on workstation includes all items noted above except one supporting column and base; this workstation shares the column and base from an existing starter workbench. Worksurfaces are available in black epoxy resin, ESD almond static-dissipative laminate with grounding kit, and wallaby plastic laminate.
Ordering information: Starter workbenches and add-on workstation do not include a work surface. Work surfaces are available separately. A starter workbench is required before purchasing an add-on workstation. Standard color is light neutral with a black accent strip; for other color options, contact your Avantor sales representative.
Spécifications
  • Désignation:
    Add-on workstation
  • Hauteur:
    36"
  • W×D:
    60×30"