AlphaMop™ Polyester Mop
AlphaMop™ polyester heads and foam pads, 10 covers and 1 pad/bag; 12 bags/case
Catalog # TW-TX7118
Supplier: Texwipe
Supplier #: TX7118
Jump to:
Product Details & Documents
Ideal for cleaning walls, floors, and other flat surfaces in critical environments.
AlphaMop™ covers TWTX7026 are pre-wetted with 6% isopropyl alcohol. The case purchase includes two foam pads. Replacement pack TW-TX7118 contains 150 polyester heads and 6 foam pads.
The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fiberglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. Lightweight mop maintains ultralow generation of particles and extractables. Bag-within-a-Bag® packaged.
Documents
Specifications
- Désignation:AlphaMop™ polyester heads and foam pads
- Packed:10 covers and 1 pad/bag; 12 bags/case