Micro-Vials, DWK Life Sciences
5.00 ml, Non
: 746100-0005
Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
- Standard taper 14/10 connections
- Ideal for use up to 500 °C
- Avec bouchon:Non
- Capacité:5.00 ml