Micro-Vials, DWK Life Sciences
3.00 ml, Non
Supplier #: 746100-0003
Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
- Standard taper 14/10 connections
- Ideal for use up to 500 °C
- Avec bouchon:Non
- Capacité:3.00 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



