Angled Two Neck Round Bottom Flask, Kimble Chase
25 ml, 42 mm, 14/20, 14/20
: 294990-0025
Distilling round bottom flask with angled side neck
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Two-neck flask with Standard Taper outer joints. Side neck is angled.
- Ø ext.:42 mm
- Capacité:25 ml
- Side ST Joint Size:14/20
- Center Joint:14/20