Angled Two Neck Round Bottom Flask, Kimble Chase
25 ml, 42 mm, 14/20, 14/20
  KT294990-0025
 :  294990-0025
Angled Two Neck Round Bottom Flask, Kimble Chase
  KT294990-0025
Angled Two Neck Round Bottom Flask, Kimble Chase
Angled Two Neck Round Bottom Flask, Kimble Chase
25 ml, 42 mm, 14/20, 14/20
$124.95
Each
Distilling round bottom flask with angled side neck
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Two-neck flask with Standard Taper outer joints. Side neck is angled.
  • Ø ext.:
    42 mm
  • Capacité:
    25 ml
  • Side ST Joint Size:
    14/20
  • Center Joint:
    14/20