Mating Factor alpha Trifluoroacetate
5 mg
H-2925.0005BA
: 4003514.0005
Mating pheromone of mating type α cells of S. cerevisiae. This factor inhibits DNA synthesis, thereby preventing cell division of mating type a cells. The hydrophobic peptide binds to a class D G-protein-coupled receptor Ste2p.
- Taille:5 mg
- Cat. No.:H-2925.0005BA
- Molecular formula:C₈₂H₁₁₄N₂₀O₁₇S
- Protein/peptide name:Mating Factor alpha Trifluoroacetate
- Storage conditions:–20 ± 5 °C
- Protein synonyms:a-Factor|Pheromone a-Factor|alpha-Factor
- MDL No.:MFCD22375527
- Sequence:H-Trp-His-Trp-Leu-Gln-Leu-Lys-Pro-Gly-Gln-Pro-Met-Tyr-OH
- Molecular weight:1684.01
- CAS No.:59401-28-4



