Cyclo(-Phe-Pro)
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
Cain et al. obtained an array of small molecule peptide mimetics targeting the melanocortin receptors from this diketopiperazine. Brauns et al. demonstrated that cyclo(Phe-Pro) inhibits the growth of HT-29, MCF-7 and HeLa cells and induces apoptosis in HT-29 colon cancer cells.
Spécifications
Spécifications
- Taille:1 g
- Cat. No.:G-4720.1000BA
- Molecular formula:C₁₄H₁₆N₂O₂
- Protein/peptide name:Cyclo(-Phe-Pro)
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038604
- Sequence:Cyclo(-Phe-Pro)
- Molecular weight:244.29
- CAS No.:3705-26-8
Spécifications
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



