Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
RIPA Lysis Buffer, Rockland

RIPA Lysis Buffer, Rockland

RIPA Lysis Buffer, 250 mL, 7.4
Catalog # CAMB-030-0250
Supplier #:  MB-030-0250
RIPA Lysis Buffer, Rockland
Catalog # CAMB-030-0250
CAS Number:  
RIPA Lysis Buffer, Rockland

RIPA Lysis Buffer, 250 mL, 7.4

CAS Number:  
Price
$439.18
Log in for availability
Each (250ml)
Minimum order quantity is 1
Product Details & Documents

RIPA (Radio-Immunoprecipitation Assay) Lysis Buffer enables rapid, efficient cell lysis and solubilization of proteins from both adherent and suspension cultured mammalian cells

In addition, RIPA Lysis Buffer minimizes non-specific protein-binding interactions to keep background low, while allowing most specific interactions to occur, enabling studies of relevant protein-protein interactions. The following RIPA Lysis Buffer components have the following effects: Tris-HCl is a buffering agent prevents protein denaturation, NaCl is a salt that prevents non-specific protein aggregation, NP-40 is a non-ionic detergent to extract proteins; Na-deoxycholate and SDS are ionic detergents to extract proteins; and sodium azide is a bacteriostatic agent added to retard bacterial growth. RIPA Lysis Buffer is supplied as a ready-to-use solution that requires no preparation. We suggest that the user add protease and phosphatase inhibitors not included with this product prior to use.

This product is ready-to-use as a working 1X solution and requires no further dilution. 1X RIPA Lysis Buffer is intended for the extraction of cellular proteins for the efficient lysis of cells and solubilization of protein, while minimizing protein degradation and maintaining protein immunoreactivity and biological activity. We recommend using 1.0 ml of RIPA Lysis Buffer to lyse 0.5 to 5 x 10E7 adherent mammalian cells. This buffer contains ionic detergents and may not be suitable for kinase enzymes, if these enzymes are easily denatured. Do not add phosphatase inhibitors when preparing lysates for phosphatase assays. 1X RIPA lysis buffer consists of 50 mM Tris HCl, 150 mM NaCl, 1.0% (v/v) NP-40, 0.5% (w/v) Sodium Deoxycholate, 1.0 mM EDTA, 0.1% (w/v) SDS and 0.01% (w/v) sodium azide at a pH of 7.4. This buffer was meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water. Protease and phosphatase inhibitors are recommended but not included in product composition.

It has long been a widely used lysis and wash buffer for small-scale affinity pull-down applications, such as immunoprecipitation, since most antibodies and protein antigens are not adversely affected by the components of this buffer.

This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.

Specifications

Specifications

  • Désignation:
    RIPA Lysis Buffer
  • Taille:
    250 mL
  • pH:
    7.4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.