Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Product Details & Documents
Vegetable based.
- GMP manufactured product
- Vegetable based
Suitable for use in the manufacture of parenteral dosage forms.
- CAS Number:9005-65-6
- Numéro MDL:MFCD00082107
- Fusion Pt:100 °C
- Densité:1.062 g/cm³
- :Management of Change (MOC) category = R
- Fusion Pt:–21 °C
- Flash Pt:113 °C (closed cup)
- Température de conservation:Température ambiante
- L'EINECS:500-019-9
Spécifications
Résultats des tests de spécification
- NF - Acid Value:≤2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤0.25%
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25 °C:1.06 - 1.09
- NF - Viscosity at 25 °C, cSt:300 - 500
- NF - Water (H₂O):≤3.0%
- NF - Peroxide Value:≤10.0
- NF - Ethylene Oxide:≤1 ppm
- NF - Dioxane:≤10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤5.0%
- NF - Composition of Fatty Acids - Palmitic Acid:≤16.0%
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤8.0%
- NF - Composition of Fatty Acids - Stearic Acid:≤6.0%
- NF - Composition of Fatty Acids - Oleic Acid:≥58.0%
- NF - Composition of Fatty Acids - Linoleic Acid:≤18.0%
- NF - Composition of Fatty Acids - Linolenic Acid:≤4.0%
- EP - Acid Value:≤2.0
- EP - Total Ash:≤0.25%
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤10.0
- EP - Ethylene Oxide:≤1 ppm
- EP - Dioxan:≤10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤3.0%
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤5.0%
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤16.0%
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤8.0%
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤6.0%
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥58.0%
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤18.0%
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤4.0%
- JP - Acid Value:≤2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤5.0%
- JP - Composition of Fatty Acids - Palmitic Acid:≤16.0%
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤8.0%
- JP - Composition of Fatty Acids - Stearic Acid:≤6.0%
- JP - Composition of Fatty Acids - Oleic Acid:≥58.0%
- JP - Composition of Fatty Acids - Linoleic Acid:≤18.0%
- JP - Composition of Fatty Acids - Linolenic Acid:≤4.0%
- JP - Dioxane:≤10 ppm
- JP - Ethylene Oxide:≤1 ppm
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤10.0
- JP - Residue on Ignition:≤0.25%
- JP - Water (H₂O):≤3.0%
- JP - Saponification Value:45 - 55
- Additional Tests - Color (Gardner):≤7
- Additional Tests - Odor (Faint):Passes Test
- Additional Tests - Water (H₂O):≤0.2%
- Additional Tests - Peroxide Value, meqO₂/kg:≤2.0
- Additional Tests - Endotoxin Concentration (EU/ml):≤10
- Free Ethylene Oxide:≤1 ppm
- 1,4-Dioxane:≤5 ppm
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤100
- Microbiological - Escherichia Coli:Passes Test
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Microbiological - Total Yeast and Mold Count (cfu/g):≤50
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol :≤ 0.10 %
Spécifications
- Type de conditionnement:Glass bottle, amber, narrow mouth
- Taille:1 L
Spécifications
Certains produits peuvent apparaître comme restreints
Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.
Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)
Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.



