Active Gauge Cables, Edwards
10 m (32 ft. 10")
Catalog # CAD400-01-100
Supplier:   Edwards
 :  D40001100
Active Gauge Cables, Edwards
Catalog # CAD400-01-100
Supplier:  Edwards
Active Gauge Cables, Edwards
10 m (32 ft. 10")
Price
$483.23
Log in pour la disponibilité
Each
Product Details & Documents
These cables connect any active gauge display or active gauge controller to any active gauge head.
Spécifications
  • Cable Length:
    10 m (32 ft. 10")