Active Gauge Cables, Edwards | Avantor
Active Gauge Cables, Edwards
5 m (16 ft. 5")
Catalog # CAD400-01-050
Supplier:   Edwards
Fournisseur #:  D40001050
Active Gauge Cables, Edwards
Catalog # CAD400-01-050
Supplier:  Edwards
Active Gauge Cables, Edwards
5 m (16 ft. 5")
Price
$325.56
Log in pour la disponibilité
Each
Product Details & Documents
These cables connect any active gauge display or active gauge controller to any active gauge head.
Spécifications
  • Cable Length:
    5 m (16 ft. 5")