Polysorbate 80
Bottle, 2.5 L
  CAAAAL13315-0F
 :  L13315-0F
Polysorbate 80
  CAAAAL13315-0F
 :  9005-65-6
Polysorbate 80
Bottle, 2.5 L
 :  9005-65-6
$276.59
Each (3l)
Used for solubilizing membrane proteins during isolation and purification
CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107
 :  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  •  :9005-65-6
  •  :MFCD00082107
  •  :14,7582
  •  :>100 °C
  •  :100 °C
  •  :1.064
  •  :113 °C (closed cup)
  •  :>149 °C(300 °F)
  •  :Température ambiante
  •  :500-019-9
  • Type de conditionnement:
    Bottle
  • Taille:
    2.5 L