ASTM Class 4 Precision Weight Sets
ASTM Class 4 Precision Weight Sets
10 mg - 50 g, 16
Catalog # CA97021-890
Supplier:   Mettler Toledo
Supplier #:  11124081
ASTM Class 4 Precision Weight Sets
Catalog # CA97021-890
Supplier:  Mettler Toledo
ASTM Class 4 Precision Weight Sets
ASTM Class 4 Precision Weight Sets
10 mg - 50 g, 16
Price
$2,640.00
Log in for availability
Each
Product Details & Documents
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
  • ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Specifications
  • Range:
    10 mg - 50 g
  • No. of Weights:
    16