ASTM Class 4 Precision Weight Sets
1 mg - 1 kg, 25
Catalog # CA97021-888
Supplier: Mettler Toledo
: 11124075
Jump to:
Product Details & Documents
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
- 316 stainless steel
- Low magnetic permeability
- High corrosion resistance
- High gloss polish
- ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Specifications
- Range:1 mg - 1 kg
- No. of Weights:25