ASTM Class 4 Precision Weight Sets
1 mg - 200 g, 23
Catalog # CA97021-884
Supplier: Mettler Toledo
Fournisseur #: 11124071
Jump to:
Product Details & Documents
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
- 316 stainless steel
- Low magnetic permeability
- High corrosion resistance
- High gloss polish
- ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Spécifications
- Range:1 mg - 200 g
- No. of Weights:23
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.