Passer à En-tête
Passer à Contenu principal
Passer à Pied de page
Avancé
Avancé
SDS
Certificats
Données du fabricant
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SCAVENGER CRTDG COL C18 PK5
Catalog # CA97013-118
Supplier:
REVVITY HEALTH SCIENCES CANADA CA
Expand image
SCAVENGER CRTDG COL C18 PK5
Catalog # CA97013-118
Supplier:
REVVITY HEALTH SCIENCES CANADA CA
Supplier Number:
2580202
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Universal Pipette Tips
Supplier: VWR International