$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SEAL HIGH PRES F/S-200 PUMP
CA97012-294
:
REVVITY HEALTH SCIENCES CANADA CA
SEAL HIGH PRES F/S-200 PUMP
CA97012-294
:
REVVITY HEALTH SCIENCES CANADA CA
:
9907345
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH