NIST Traceable pH Buffers with Temperature Table
Bottle, 2×500 ml Each pH, 4/7/10
  CA97007-790
 :  63056116
NIST Traceable pH Buffers with Temperature Table
  CA97007-790
 :  
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table
Bottle, 2×500 ml Each pH, 4/7/10
 :  
$279.00
Pack of 6 (500ml)
NIST-traceable buffer for pH electrode calibration and verification.
  • Temperature table on bottle
Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.
  • Type de conditionnement:
    Bottle
  • Taille:
    2×500 ml Each pH
  • pH:
    4/7/10