NIST Traceable pH Buffers with Temperature Table
Bottle, 2×500 ml Each pH, 4/7/10
: 63056116
NIST-traceable buffer for pH electrode calibration and verification.
- Temperature table on bottle
Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.
- Type de conditionnement:Bottle
- Taille:2×500 ml Each pH
- pH:4/7/10