Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
NIST Traceable pH Buffers with Temperature Table | Avantor

NIST Traceable pH Buffers with Temperature Table

Bottle, 500 ml, 7.00
Catalog # CA97007-786
Supplier:   Mettler Toledo
Fournisseur #:  63056125
NIST Traceable pH Buffers with Temperature Table
Catalog # CA97007-786
Supplier:  Mettler Toledo
CAS Number:  
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table

Bottle, 500 ml, 7.00

CAS Number:  
Price
$36.30
Log in pour la disponibilité
Each (500ml)
Product Details & Documents

NIST-traceable buffer for pH electrode calibration and verification.

  • Temperature table on bottle

Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.

Documents

Spécifications

Spécifications

  • Type de conditionnement:
    Bottle
  • Taille:
    500 ml
  • pH:
    7.00

Spécifications