Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Electrode Fill Solutions | Avantor

Electrode Fill Solutions

1M KNO₃, Bottle, 25 ml, Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide
Catalog # CA97001-718
Supplier:   Mettler Toledo
Fournisseur #:  51343182
Electrode Fill Solutions
Catalog # CA97001-718
Supplier:  Mettler Toledo
CAS Number:  
Electrode Fill Solutions
Electrode Fill Solutions

1M KNO₃, Bottle, 25 ml, Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide

CAS Number:  
Price
$34.50
Log in pour la disponibilité
Each (25ml)
Product Details & Documents

FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.

Delivery information: Supplied in 25 ml (0.9 oz.) bottles.

Documents

Spécifications

Spécifications

  • Type de conditionnement:
    Bottle
  • Désignation:
    1M KNO₃
  • Ion:
    Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide
  • Volume:
    25 ml

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.