Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mag-Bind SeqDTR, Omega Bio-Tek

Mag-Bind SeqDTR, Omega Bio-Tek

Mag-Bind SeqDTR, 500 mL
Catalog # CA95059-688
Supplier:   Omega Bio-Tek
Supplier #:  M1300-50
Mag-Bind SeqDTR, Omega Bio-Tek
Catalog # CA95059-688
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind SeqDTR, Omega Bio-Tek

Mag-Bind SeqDTR, 500 mL

CAS Number:  
Price
$21,759.81
Log in pour la disponibilité
Each (500ml)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Omega Bio-teks Mag-Bind SeqDTR is designed to effective and reliable removal of unincorporated terminators from sequencing reaction. The system combines Omega Bio-teks proprietary chemistry with the reversible nucleic acid-binding properties of paramagnetic beads to eliminate excess nucleotides, primers, salts, and unicorporated dye terminators.

  • Long Phred 20 read lengths averaging over 800 bps
  • Works with automated liquid handlers
  • Can be used in 96 or 384 format

This kit is designed for both manual and fully automated purification of sequencing products, The chemistry has been successfully automated on liquid handlers such has Beckman Coulter’s Biomek FX/NX, and Hamilton STAR/Starlet. Mag-Bind SEQ DTR high recovery rate allows for decreased usage of ABI Big Dye chemistry, normally 1/16 or 1/32 reactions can be performed without a decrease in signal strength when compared to sephadex based clean-up

Spécifications

Spécifications

  • Désignation:
    Mag-Bind SeqDTR
  • Taille:
    500 mL
  • Cat. No.:
    CA95059-688
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Description:
    Mag-Bind SeqDTR
  • Sample type:
    PCR and other enzymatic reactions

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.