Colorant selon Wright
1 gal.
Supplier: Azer Scientific
Fournisseur #: ES920
Jump to:
Product Details & Documents
Wright stain is a polychromatic hematology stain consisting of eosin and methylene blue.
- Dye mixture produces multiple colors to clearly differentiate white blood cells, red blood cells appear red-orange
- Suitable for automated and manual staining protocols
- Wright stain can be used for urine cytology and fine needle aspirate cell preparations
Synonymes: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
- CAS Number:68988-92-1
- Numéro MDL:MFCD00082143
Spécifications
Spécifications
- Taille:1 gal.



