Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Colorant selon Wright

Colorant selon Wright

32 oz.
Catalog # CA95057-508
Supplier:   Azer Scientific
Supplier #:  ES918
Colorant selon Wright
Catalog # CA95057-508
Supplier:  Azer Scientific
CAS Number:  68988-92-1
Colorant selon Wright

32 oz.

CAS Number:  68988-92-1
Price
$1,299.60
Log in for availability
Each (32oz)
Product Details & Documents

Wright stain is a polychromatic hematology stain consisting of eosin and methylene blue.

  • Dye mixture produces multiple colors to clearly differentiate white blood cells, red blood cells appear red-orange
  • Suitable for automated and manual staining protocols
  • Wright stain can be used for urine cytology and fine needle aspirate cell preparations
Synonyms:  Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
  • CAS Number:68988-92-1
  • MDL Number:MFCD00082143
Specifications

Specifications

  • Taille:
    32 oz.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.