SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Biopsy Cassettes, Blue
Catalog # CA95030-266
Supplier:   Simport Scientific
Supplier #:  M510-6SL
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Catalog # CA95030-266
Supplier:  Simport Scientific
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Biopsy Cassettes, Blue
Price
$615.41
Log in pour la disponibilité
Case of 750
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. The compact design features a space-saving recessed cover. The one-piece integral lid eliminates the need for separate steel lids. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle. All cassettes colors are non-cytotoxic and non-metallic. Dimensions: 41Wx28.5Lx6Hmm (15/8x11/8x1/4").
 : Cassettes are packaged in transparent sleeves for quick loading into automatic labeling instruments. Each case contains 10 sleeves of 75 cassettes.
Spécifications
  • Color:
    Blue
  • Désignation:
    Biopsy Cassettes