Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific

SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific

Tissue Cassettes, White
Catalog # CA95030-230
Supplier:   Simport Scientific
Supplier #:  M509-2SL
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Catalog # CA95030-230
Supplier:  Simport Scientific
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
SLIMSETTE™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific

Tissue Cassettes, White

Price
$615.41
Log in for availability
Case of 750
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid

The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. The compact design features a space-saving recessed cover. The one-piece integral lid eliminates the need for separate steel lids. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle. All cassettes colors are non-cytotoxic and non-metallic. Dimensions: 41Wx28.5Lx6Hmm (15/8x11/8x1/4").

Ordering information: Cassettes are packaged in transparent sleeves for quick loading into automatic labeling instruments. Each case contains 10 sleeves of 75 cassettes.

Specifications
  • Color:
    White
  • Désignation:
    Tissue Cassettes

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.