Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BBL™ Thioglycollate Medium without Indicator, BD Life Sciences | Avantor

BBL™ Thioglycollate Medium without Indicator, BD Life Sciences

BBL™ Thioglycollate Medium without Indicator, 500 g
Catalog # CA90000-298
Supplier:   BD
Fournisseur #:  211720
BBL™ Thioglycollate Medium without Indicator, BD Life Sciences
Catalog # CA90000-298
Supplier:  BD
CAS Number:  
BBL™ Thioglycollate Medium without Indicator, BD Life Sciences
BBL™ Thioglycollate Medium without Indicator, BD Life Sciences

BBL™ Thioglycollate Medium without Indicator, 500 g

CAS Number:  
Price
292,00 $
Log in pour la disponibilité
Each (500g)
Product Details & Documents

Thioglycollate Medium without Indicator is a general-purpose medium for the cultivation of microorganisms, especially obligate anaerobes.

  • Casein and soybean peptones, dextrose, and cystine supply nitrogen and carbon
  • Sodium chloride provides essential ions

Casein and soybean peptones, dextrose, and cystine supply nitrogenous and carbonaceous compounds, fermentable carbohydrates, and trace ingredients. This medium is satisfactorily used as an enrichment culture for various types of specimens and also as a transportation medium.

Spécifications

Spécifications

  • Désignation:
    BBL™ Thioglycollate Medium without Indicator
  • Taille:
    500 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.