Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cable Attachments, Brady Worldwide®

Cable Attachments, Brady Worldwide®

Nylon cable attachment, Red, 2.4 m (8 ft.)
Catalog # CA89007-224
Supplier:   Brady Worldwide
Supplier #:  50932
Cable Attachments, Brady Worldwide®
Catalog # CA89007-224
Supplier:  Brady Worldwide
Cable Attachments, Brady Worldwide®
Cable Attachments, Brady Worldwide®

Nylon cable attachment, Red, 2.4 m (8 ft.)

Price
$34.89
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Cable attachments are designed to lock out gate valves

Cable attachments are available in two formats. Choose between a 1/8" nonconductive nylon cable or a rustproof sheathed metal cable.

Specifications
  • Color:
    Red
  • Désignation:
    Nylon cable attachment
  • Longueur:
    2.4 m (8 ft.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.