Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences

Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences

Optimizer Buffer™-VI [5X], Glyoxal groups, Amine
Catalog # CA82022-964
Supplier:   G-Biosciences
Supplier #:  BKC-09
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
Catalog # CA82022-964
Supplier:  G-Biosciences
CAS Number:  
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences

Optimizer Buffer™-VI [5X], Glyoxal groups, Amine

CAS Number:  
Price
$551.53
Log in pour la disponibilité
Pack of 2 (25ml)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The conjugation and modification reactions used to cross-link proteins or couple labels to proteins, such as biotin, enzymes, and fluorescent dyes, require certain conditions, including pH and chemical composition, for optimal conjugation

G-Biosciences has prepared six reaction-specific buffers that provide the optimal conditions for protein labeling, modification, and cross reaction. Optimizer Buffer™ -I is free of amines and is specific for amine reactive groups, such as imidoesters and N-hydroxysuccinimide (NHS) esters. Optimizer Buffer™-II has a high pH and is specifically designed for the sulfhydryl reactive haloacetyls, such as iodoacetyl. Optimizer Buffer™-III is designed for sulfhydryl reactive groups, maleimides and pyridyl disulfides, and has reagents that prevent oxidation of free sulfhydryls. Optimizer Buffer™-IV is designed to provide the optimal conditions necessary for carboxyl reactive reagents. Optimizer Buffer™-V is a modified acetate buffer that is specific for carbohydrate reactive groups. Optimizer Buffer™-VI is an alkaline buffer specific for the formation of Schiff’s base between amines and glyoxal groups.

G-Biosciences' buffers contain optimized concentrations of buffering agents, pH and other cofactors for specific reactions. Simply exchange the buffer of your sample with a suitable Optimizer Buffer™, and you are ready for efficient reaction. Use of G-Biosciences' SpinOUT™ or Tube-O-DIALYZER™ is recommended for buffer exchange and optimal reaction results. Each Optimizer Buffer™ is supplied as a 5X concentrated buffer.

Many common buffers routinely used in laboratories have an inhibitory effect on conjugation reactions; for example, Tris buffers inhibit coupling to amines.

Spécifications

Spécifications

  • Désignation:
    Optimizer Buffer™-VI [5X]
  • Reactive Group:
    Glyoxal groups
  • Target Group:
    Amine

Spécifications