Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences

Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences

Optimizer Buffer™-II [5X], Iodoacetyl groups, Sulfhydryl
Catalog # CA82022-956
Supplier:   G-Biosciences
Supplier #:  BKC-05
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
Catalog # CA82022-956
Supplier:  G-Biosciences
CAS Number:  
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences

Optimizer Buffer™-II [5X], Iodoacetyl groups, Sulfhydryl

CAS Number:  
Price
$551.53
Log in for availability
Pack of 2 (25ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The conjugation and modification reactions used to cross-link proteins or couple labels to proteins, such as biotin, enzymes, and fluorescent dyes, require certain conditions, including pH and chemical composition, for optimal conjugation

G-Biosciences has prepared six reaction-specific buffers that provide the optimal conditions for protein labeling, modification, and cross reaction. Optimizer Buffer™ -I is free of amines and is specific for amine reactive groups, such as imidoesters and N-hydroxysuccinimide (NHS) esters. Optimizer Buffer™-II has a high pH and is specifically designed for the sulfhydryl reactive haloacetyls, such as iodoacetyl. Optimizer Buffer™-III is designed for sulfhydryl reactive groups, maleimides and pyridyl disulfides, and has reagents that prevent oxidation of free sulfhydryls. Optimizer Buffer™-IV is designed to provide the optimal conditions necessary for carboxyl reactive reagents. Optimizer Buffer™-V is a modified acetate buffer that is specific for carbohydrate reactive groups. Optimizer Buffer™-VI is an alkaline buffer specific for the formation of Schiff’s base between amines and glyoxal groups.

G-Biosciences' buffers contain optimized concentrations of buffering agents, pH and other cofactors for specific reactions. Simply exchange the buffer of your sample with a suitable Optimizer Buffer™, and you are ready for efficient reaction. Use of G-Biosciences' SpinOUT™ or Tube-O-DIALYZER™ is recommended for buffer exchange and optimal reaction results. Each Optimizer Buffer™ is supplied as a 5X concentrated buffer.

Many common buffers routinely used in laboratories have an inhibitory effect on conjugation reactions; for example, Tris buffers inhibit coupling to amines.

Specifications

Specifications

  • Désignation:
    Optimizer Buffer™-II [5X]
  • Reactive Group:
    Iodoacetyl groups
  • Target Group:
    Sulfhydryl

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.