G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution) | Avantor
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl), 1 gal.
Catalog # CA82021-482
Supplier:   G-Biosciences
Fournisseur #:  R018
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Catalog # CA82021-482
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl), 1 gal.
CAS Number:  
Price
$625.94
Log in pour la disponibilité
Each (4l)
La quantité minimale de commande est de 1
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1 M Tris·HCl (pH 7.4)
  • 1.5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Spécifications
Spécifications
  • Désignation:
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)
  • Taille:
    1 gal.