G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl), 1 gal.
Catalog # CA82021-482
Supplier: G-Biosciences
Fournisseur #: R018
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Catalog # CA82021-482
Supplier: G-Biosciences
CAS Number:
Jump to:
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
1 M Tris·HCl (pH 7.4)
- 1.5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Spécifications
Spécifications
- Désignation:Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)
- Taille:1 gal.
Spécifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.